Anti-HIST1H2BN Rabbit Polyclonal Antibody

Lieferant: Abgent
AP13115A
ABGEAP13115AEA 354 EUR
ABGEAP13115A
Anti-HIST1H2BN Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HIST1H2BN (histone cluster 1, H2bn)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR