Anti-DNAJC14 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP13146C
ABGEAP13146CEA 354 EUR
ABGEAP13146C
Anti-DNAJC14 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DNAJC14 (DnaJ (Hsp40) homolog, subfamily C, member 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR