Anti-TGFBRAP1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP13395B
ABGEAP13395BEA 354 EUR
ABGEAP13395B
Anti-TGFBRAP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TGFBRAP1 (transforming growth factor, beta receptor associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR