Anti-CD5 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP13533C
ABGEAP13533CEA 354 EUR
ABGEAP13533C
Anti-CD5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CD5 (CD5 molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR