Anti-TRIM23 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP13641A
ABGEAP13641AEA 354 EUR
ABGEAP13641A
Anti-TRIM23 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TRIM23 (tripartite motif containing 23)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR