Anti-PPP1R3F Rabbit Polyclonal Antibody

Lieferant: Abgent
AP13676B
ABGEAP13676BEA 354 EUR
ABGEAP13676B
Anti-PPP1R3F Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PPP1R3F (protein phosphatase 1, regulatory subunit 3F)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR