Anti-H1FX Rabbit Polyclonal Antibody

Lieferant: Abgent
AP13731B
ABGEAP13731BEA 354 EUR
ABGEAP13731B
Anti-H1FX Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: H1FX (H1 histone family, member X)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR