Anti-OR1N1 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP13812B
ABGEAP13812BEA 354 EUR
ABGEAP13812B
Anti-OR1N1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OR1N1 (olfactory receptor, family 1, subfamily N, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human