Anti-UGT1A1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP13949A
ABGEAP13949AEA 354 EUR
ABGEAP13949A
Anti-UGT1A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UGT1A1 (UDP glucuronosyltransferase 1 family, polypeptide A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR