Anti-Pak7 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP14074A
ABGEAP14074AEA 354 EUR
ABGEAP14074A
Anti-Pak7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Pak7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR