Anti-USP46 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP14161B
ABGEAP14161BEA 354 EUR
ABGEAP14161B
Anti-USP46 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: USP46 (ubiquitin specific peptidase 46)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR