Anti-MAP3K3 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP14251C
ABGEAP14251CEA 354 EUR
ABGEAP14251C
Anti-MAP3K3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAP3K3 (mitogen-activated protein kinase kinase kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Mouse