Anti-DEFB112 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP14332A
ABGEAP14332AEA 354 EUR
ABGEAP14332A
Anti-DEFB112 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DEFB112 (defensin, beta 112)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR