Anti-BCAS3 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP1437B
ABGEAP1437BEA 354 EUR
ABGEAP1437B
Anti-BCAS3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BCAS3 (breast carcinoma amplified sequence 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human