Anti-PPM1G Rabbit Polyclonal Antibody

Lieferant: Abgent
AP14396A
ABGEAP14396AEA 354 EUR
ABGEAP14396A
Anti-PPM1G Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PPM1G (protein phosphatase, Mg2+/Mn2+ dependent, 1G)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR