Anti-STS Rabbit Polyclonal Antibody
Lieferant: Abgent
AP14527B
ABGEAP14527BEA 354 EUR
ABGEAP14527B
Anti-STS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STS (steroid sulfatase (microsomal), isozyme S)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human