Anti-RING1 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP14560A
ABGEAP14560AEA 354 EUR
ABGEAP14560A
Anti-RING1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RING1 (ring finger protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human