Anti-U2AF2 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP14583A
ABGEAP14583AEA 354 EUR
ABGEAP14583A
Anti-U2AF2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: U2AF2 (U2 small nuclear RNA auxiliary factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human