Anti-ADCK1 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP14607B
ABGEAP14607BEA 354 EUR
ABGEAP14607B
Anti-ADCK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADCK1 (aarF domain containing kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse