Anti-DDR2 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP14616B
ABGEAP14616BEA 354 EUR
ABGEAP14616B
Anti-DDR2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DDR2 (discoidin domain receptor tyrosine kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR