Anti-VAMP3 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP14660C
ABGEAP14660CEA 354 EUR
ABGEAP14660C
Anti-VAMP3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: VAMP3 (vesicle-associated membrane protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR