Anti-TEC Rabbit Polyclonal Antibody
Lieferant: Abgent
AP14777C
ABGEAP14777CEA 354 EUR
ABGEAP14777C
Anti-TEC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TEC (tec protein tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Mouse