Anti-NUDC Rabbit Polyclonal Antibody

Lieferant: Abgent
AP16056C
ABGEAP16056CEA 354 EUR
ABGEAP16056C
Anti-NUDC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NUDC (nudC nuclear distribution protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR