Anti-MAP2K6 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP16108A
ABGEAP16108AEA 354 EUR
ABGEAP16108A
Anti-MAP2K6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAP2K6 (mitogen-activated protein kinase kinase 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse