Anti-KRT28 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP16179B
ABGEAP16179BEA 354 EUR
ABGEAP16179B
Anti-KRT28 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KRT28 (keratin 28)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR