Anti-RPL12 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP16275C
ABGEAP16275CEA 354 EUR
ABGEAP16275C
Anti-RPL12 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RPL12 (ribosomal protein L12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR