Anti-TP53BP2 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP16353A
ABGEAP16353AEA 354 EUR
ABGEAP16353A
Anti-TP53BP2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TP53BP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR