Anti-ZNF436 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP16509A
ABGEAP16509AEA 354 EUR
ABGEAP16509A
Anti-ZNF436 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF436
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human