Anti-TJP3 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP16653A
ABGEAP16653AEA 354 EUR
ABGEAP16653A
Anti-TJP3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TJP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR