Anti-IQGAP1 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP16889A
ABGEAP16889AEA 354 EUR
ABGEAP16889A
Anti-IQGAP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IQGAP1 (IQ motif containing GTPase activating protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human