Anti-PAF1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP17025B
ABGEAP17025BEA 354 EUR
ABGEAP17025B
Anti-PAF1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PAF1 (Paf1, RNA polymerase II associated factor, homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR