Anti-SNRNP70 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP17045C
ABGEAP17045CEA 354 EUR
ABGEAP17045C
Anti-SNRNP70 Rabbit Polyclonal Antibody
Antikörper
LEAF (Low Endotoxin, Azide-Free)

Western Blot: 1:1000

Type: Primary
Antigen: SNRNP70 (small nuclear ribonucleoprotein 70kDa (U1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR