Anti-RRM2B Rabbit Polyclonal Antibody

Lieferant: Abgent
AP17054C
ABGEAP17054CEA 354 EUR
ABGEAP17054C
Anti-RRM2B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RRM2B (ribonucleotide reductase M2 B (TP53 inducible))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR