Anti-BRCA1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP17140A
ABGEAP17140AEA 354 EUR
ABGEAP17140A
Anti-BRCA1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR