Anti-QPCT Rabbit Polyclonal Antibody

Lieferant: Abgent
AP17434B
ABGEAP17434BEA 354 EUR
ABGEAP17434B
Anti-QPCT Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: QPCT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR