Anti-AIDA Rabbit Polyclonal Antibody

Lieferant: Abgent
AP17627B
ABGEAP17627BEA 354 EUR
ABGEAP17627B
Anti-AIDA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AIDA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR