Anti-OR2L8 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP17638B
ABGEAP17638BEA 354 EUR
ABGEAP17638B
Anti-OR2L8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OR2L8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR