Anti-LZIC Rabbit Polyclonal Antibody
Lieferant: Abgent
AP17735C
ABGEAP17735CEA 354 EUR
ABGEAP17735C
Anti-LZIC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LZIC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human