Anti-MSL3 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP17736A
ABGEAP17736AEA 354 EUR
ABGEAP17736A
Anti-MSL3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MSL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human