Anti-NXNL1 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP17740C
ABGEAP17740CEA 354 EUR
ABGEAP17740C
Anti-NXNL1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NXNL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human