Anti-PPIL6 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP17743A
ABGEAP17743AEA 354 EUR
ABGEAP17743A
Anti-PPIL6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PPIL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human