Anti-ZNF565 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP17944A
ABGEAP17944AEA 354 EUR
ABGEAP17944A
Anti-ZNF565 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF565
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human