Anti-DFNB59 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP18071C
ABGEAP18071CEA 354 EUR
ABGEAP18071C
Anti-DFNB59 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DFNB59
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human