Anti-ATG3 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP1807B
ABGEAP1807BEA 354 EUR
ABGEAP1807B
Anti-ATG3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ATG3 (autophagy related 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR