Anti-MS4A8 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP18101A
ABGEAP18101AEA 354 EUR
ABGEAP18101A
Anti-MS4A8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MS4A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human