Anti-OR51B6 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP18220B
ABGEAP18220BEA 354 EUR
ABGEAP18220B
Anti-OR51B6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OR51B6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human