Anti-OSTM1 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP18345B
ABGEAP18345BEA 354 EUR
ABGEAP18345B
Anti-OSTM1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OSTM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human