Anti-HOXA9 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP18401C
ABGEAP18401CEA 354 EUR
ABGEAP18401C
Anti-HOXA9 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HOXA9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR