Anti-SSX5 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP18520A
ABGEAP18520AEA 354 EUR
ABGEAP18520A
Anti-SSX5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SSX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR