Anti-MCF2L Rabbit Polyclonal Antibody

Lieferant: Abgent
AP18793B
ABGEAP18793BEA 354 EUR
ABGEAP18793B
Anti-MCF2L Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MCF2L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR